2',6'-Difluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2,6-Difluorophenyl)propan-1-one

004437-1G 004437-5G
101787-246EA 30.77 USD
101787-246 101787-248
2',6'-Difluoropropiophenone ≥97%
2',6'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Boiling Pt: 71 °C (4mm)
MDL Number: MFCD00015506
CAS Number: 85068-31-1
EINECS: 285-293-5

Order Now

Specification Test Results

Boiling point 71°C/4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR