3',4'-Difluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3,4-Difluorophenyl)propan-1-one

004436-1G 004436-5G
101787-242EA 24.62 USD
101787-242 101787-244
3',4'-Difluoropropiophenone ≥97%
3',4'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Boiling Pt: 65 °C (1.8mm)
MDL Number: MFCD00015505
CAS Number: 23384-72-7
EINECS: 245-627-2

Order Now

Specification Test Results

Boiling point 65°C/1.8mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR