2',4'-Difluoropropiophenone ≥95%

Supplier: Matrix Scientific

Synonyms: 1-(2,4-Difluorophenyl)propan-1-one

004435-5G 004435-25G
101787-238EA 24.62 USD
101787-238 101787-240
2',4'-Difluoropropiophenone ≥95%
2',4'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Boiling Pt: 52 °C (1013 hPa)
Density: 1.193 g/cm³ (20 °C)
MDL Number: MFCD00015507
CAS Number: 85068-30-0
EINECS: 285-291-4

Order Now

Specification Test Results

Boiling point 51-52°C/1.5
ND20 1.485

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR