2',5'-Difluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2,5-Difluorophenyl)propan-1-one

004434-1G 004434-5G
101787-234EA 32.31 USD
101787-234 101787-236
2',5'-Difluoropropiophenone ≥97%
2',5'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Boiling Pt: 39…40 °C (0.5mm)
MDL Number: MFCD00015509
CAS Number: 29112-90-1
EINECS: 249-445-4

Order Now

Specification Test Results

Boiling point 39-40°C/0.5

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR