3',5'-Difluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3,5-Difluorophenyl)propan-1-one

004430-5G 004430-25G
101787-230EA 169.23 USD
101787-230 101787-232
3',5'-Difluoropropiophenone ≥97%
3',5'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.16 g/mol
Melting Pt: 25…27 °C
MDL Number: MFCD00061142
CAS Number: 135306-45-5

Order Now

Specification Test Results

Melting point 25-27°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR