2',3'-Difluoropropiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(2,3-Difluorophenyl)propan-1-one

004429-1G 004429-5G
101787-226EA 40 USD
101787-226 101787-228
2',3'-Difluoropropiophenone ≥97%
2',3'-Difluoropropiophenone

Hazard code(s): Irritant

Formula: C₉H₈F₂O
MW: 170.156 g/mol
MDL Number: MFCD00061141
CAS Number: 236754-63-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR