1-(3-(Trifluoromethyl)phenyl)piperazine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-(Trifluoromethyl)-N-phenylpiperazine

004406-5G 004406-25G 004406-100G
101787-184EA 193.85 USD
101787-184 101787-186 101787-188
1-(3-(Trifluoromethyl)phenyl)piperazine ≥97%
1-(3-(Trifluoromethyl)phenyl)piperazine

Hazard code(s): Irritant

Formula: C₁₁H₁₃F₃N₂
Density: 1.250 g/cm³
MDL Number: MFCD00005959
CAS Number: 15532-75-9
EINECS: 239-574-4

Order Now

Specification Test Results

Density 1.25

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR