Ethyl-3-fluorobenzoate ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-fluorobenzoate, 3-Fluorobenzoic acid ethyl ester
004394-25G
004394-100G
101787-170EA
30.77
USD
101787-170
101787-172
Ethyl-3-fluorobenzoate ≥98% for synthesis
Ethyl-3-fluorobenzoate
Hazard code(s): Irritant
Formula:
C₉H₉FO₂ Boiling Pt: 94…95 °C (16 mm) Hg Density: 1.136 g/cm³ |
MDL Number:
MFCD00000336 CAS Number: 451-02-5 EINECS: 207-191-1 |
Specification Test Results
Boiling point | 94-95°C/16 |
Density | 1.136 |
ND20 | 1.485 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...