Ethyl-2-fluorobenzoate ≥98%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-fluorobenzoate, 2-Fluorobenzoic acid ethyl ester
004393-25G
004393-100G
101787-166EA
23.08
USD
101787-166
101787-168
Ethyl-2-fluorobenzoate ≥98%
Ethyl-2-fluorobenzoate
Hazard code(s): Irritant
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Boiling Pt: 115 °C (30mm) |
MDL Number:
MFCD00039214 CAS Number: 443-26-5 EINECS: 207-134-0 |
Specification Test Results
Boiling point | 115°C/30mm |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...