3',5'-Difluoroacetophenone ≥97%

Supplier: Matrix Scientific

Synonyms: 1-(3,5-Difluorophenyl)ethan-1-one

004391-5G 004391-100G
101787-160EA 193.85 USD
101787-160 101787-164
3',5'-Difluoroacetophenone ≥97%
3',5'-Difluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 94…95 °C (13mm)
Melting Pt: 35…38 °C
MDL Number: MFCD00042489
CAS Number: 123577-99-1

Order Now

Specification Test Results

Boiling point 94-95°C/13m
Melting point 35-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR