2',6'-Difluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2,6-Difluorophenyl)ethan-1-one, 1-(2,6-Difluorophenyl)ethanone

004387-5G 004387-25G 004387-100G
101787-144EA 38.46 USD
101787-144 101787-146 101787-148
2',6'-Difluoroacetophenone ≥98%
2',6'-Difluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 76…79 °C (15mm)
Density: 1.19 g/cm³ (20 °C)
MDL Number: MFCD00000328
CAS Number: 13670-99-0
EINECS: 237-151-9

Order Now

Specification Test Results

Boiling point 76-79°C/15m
Density 1.197
ND20 1.48

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR