2',5'-Difluoroacetophenone ≥98%

Supplier: Matrix Scientific

Synonyms: 1-(2,5-Difluorophenyl)ethanone, 1-(2,5-Difluorophenyl)ethan-1-one

004383-5G 004383-25G 004383-100G
101787-130EA 21.54 USD
101787-130 101787-132 101787-134
2',5'-Difluoroacetophenone ≥98%
2',5'-Difluoroacetophenone

Hazard code(s): Irritant

Formula: C₈H₆F₂O
MW: 156.13 g/mol
Boiling Pt: 70…80 °C (15 mmHg)
Density: 1.2099 g/cm³ (22 °C)
MDL Number: MFCD00009898
CAS Number: 1979-36-8
EINECS: 217-837-4

Order Now

Specification Test Results

Density 1.233
ND20 1.4882

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR