Bis(phenylthio)methane ≥97% for synthesis

Supplier: Matrix Scientific

004354-5G 004354-25G
101787-106EA 87.69 USD
101787-106 101787-108
Bis(phenylthio)methane ≥97% for synthesis
Bis(phenylthio)methane

Hazard code(s): Stench

Formula: C₁₃H₁₂S₂
Boiling Pt: 194 °C (8mm)
Melting Pt: 36…38 °C
MDL Number: MFCD00003067
CAS Number: 3561-67-9
EINECS: 222-624-4

Order Now

Specification Test Results

Boiling point 194°C/8mm
Melting point 36-38°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR