O-p-Tolyl chlorothioformate

Supplier: Matrix Scientific

Synonyms: O-4-Methylphenyl chlorothioformate, p-Tolyl chlorothionoformate

004346-5G 004346-25G
101787-098EA 83.08 USD
101787-098 101787-100
O-p-Tolyl chlorothioformate
O-p-Tolyl chlorothioformate

Hazard code(s): Corrosive, Stored under Argon

Formula: C₈H₇ClOS
MW: 186.66 g/mol
Boiling Pt: 244…246 °C
MDL Number: MFCD00014466
CAS Number: 937-63-3
EINECS: 213-333-3
UN: 3265
ADR: 8 II

Order Now

Specification Test Results

Boiling point 52-53°C/0.1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR