3-(Diethylamino)propyl isothiocyanate ≥98%

Supplier: Matrix Scientific

004275-1G 004275-5G
101787-042EA 16.92 USD
101787-042 101787-044
3-(Diethylamino)propyl isothiocyanate ≥98%
3-(Diethylamino)propyl isothiocyanate

Hazard code(s): Lachrymator, Refrigerate

Formula: C₈H₁₆N₂S
Boiling Pt: 88 - 92 °C (0.1 -rr)
Density: 0.9535 g/cm³ (20 °C)
MDL Number: MFCD00041133
CAS Number: 2626-52-0

Order Now

Specification Test Results

Boiling point 94-95°C/3.5
Density 0.95
ND20 1.503


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR