Cyclohexyl anthranilate ≥98%

Supplier: Matrix Scientific

004255-5G 004255-25G
101787-020EA 83.08 USD
101787-020 101787-022
Cyclohexyl anthranilate ≥98%
Cyclohexyl anthranilate

Hazard code(s): Irritant

Formula: C₁₃H₁₇NO₂
MW: 219.28 g/mol
Boiling Pt: 134…137 °C (2mm)
Melting Pt: 135 °C
Density: 1.117 g/cm³ (20 °C)
MDL Number: MFCD00036492
CAS Number: 7779-16-0
EINECS: 231-920-2

Order Now

Specification Test Results

Boiling point 134-137°C/2
Density 1.11

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR