2-Fluoromesitylene ≥99%

Supplier: Matrix Scientific

Synonyms: 2-Fluoro-1,3,5-trimethylbenzene, Fluoromesitylene, 2,4,6-Trimethylfluorobenzene

004246-1G 004246-5G
101787-012EA 69.23 USD
101787-012 101787-014
2-Fluoromesitylene ≥99%
2-Fluoromesitylene

Hazard code(s): Irritant

Formula: C₉H₁₁F
MW: 138.18 g/mol
Boiling Pt: 70 °C (25mm)
Density: 0.9745 g/cm³
MDL Number: MFCD00052365
CAS Number: 392-69-8

Order Now

Specification Test Results

Boiling point 70°C/25mm
Density 0.9745
ND20 1.484

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR