(4-Nitrophenyl)acetic acid ethyl ester ≥98%

Supplier: Matrix Scientific

004245-5G 004245-25G
101787-008EA 80 USD
101787-008 101787-010
(4-Nitrophenyl)acetic acid ethyl ester ≥98%
(4-Nitrophenyl)acetic acid ethyl ester

Hazard code(s): Irritant

Formula: C₁₀H₁₁NO₄
Melting Pt: 64…66 °C
MDL Number: MFCD00017046
CAS Number: 5445-26-1
EINECS: 226-646-5

Order Now

Specification Test Results

Melting point 64-66°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR