4-Chlorophenyl phenyl ether ≥99% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Chlorodiphenyl ether, 1-Chloro-4-phenoxybenzene

004220-5G 004220-25G 004220-100G
101786-976EA 26.15 USD
101786-976 101786-978 101786-980
4-Chlorophenyl phenyl ether ≥99% for synthesis
4-Chlorophenyl phenyl ether

Hazard code(s): Irritant

Formula: C₁₂H₉ClO
MW: 204.66
Boiling Pt: 295
Melting Pt: -8
Density: 1.193 g/cm³
MDL Number: MFCD00055431
CAS Number: 7005-72-3
EINECS: 230-281-7

Order Now

Specification Test Results

Boiling point 161-162°C/1
Density 1.193
ND20 1.587


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR