1-Fluoro-4-phenoxybenzene ≥99%
Supplier: Matrix Scientific
004219-5G
004219-25G
101786-972EA
50.77
USD
101786-972
101786-974
1-Fluoro-4-phenoxybenzene ≥99%
1-Fluoro-4-phenoxybenzene
Hazard code(s): Irritant
Formula:
C₁₂H₉FO MW: 188.2 g/mol Boiling Pt: 147…149 °C (53mm) Density: 1.14 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00055239 CAS Number: 330-84-7 EINECS: 206-357-0 |
Specification Test Results
Boiling point | 147-149°C/5 |
Density | 1.14 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...