4-Fluorobiphenyl ≥97%

Supplier: Matrix Scientific

Synonyms: PFB No. 3, 4-Fluoro-1,1-biphenyl

004216-5G 004216-25G
101786-968EA 53.85 USD
101786-968 101786-970
4-Fluorobiphenyl ≥97%
4-Fluorobiphenyl

Hazard code(s): Irritant

Formula: C₁₂H₉F
MW: 172.2 g/mol
Boiling Pt: 253 °C (1013 hPa)
Melting Pt: 59 - 61 °C
Density: 1.247 g/cm³ (25 °C)
MDL Number: MFCD00011650
CAS Number: 324-74-3
EINECS: 206-304-1

Order Now

Specification Test Results

Boiling point 310°C
Melting point 74-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR