5-Bromo-2-thenoic acid ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Bromothiophene-2-carboxylic acid, 5-Bromo-2-thiophenecarboxylic acid

004204-5G 004204-25G
101786-950EA 32.31 USD
101786-950 101786-952
5-Bromo-2-thenoic acid ≥97% for synthesis
5-Bromo-2-thenoic acid

Hazard code(s): Irritant

Formula: C₅H₃BrO₂S
Melting Pt: 145…147 °C
MDL Number: MFCD00079725
CAS Number: 7311-63-9

Order Now

Specification Test Results

Melting point 145-147°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR