2-Bromothioanisole ≥99%

Supplier: Matrix Scientific
Warning

004191-5G 004191-25G
101786-922EA 15.38 USD
101786-922 101786-924
2-Bromothioanisole ≥99%
2-Bromothioanisole

Hazard code(s): Irritant, Stench

Formula: C₇H₇BrS
Boiling Pt: 145…146 °C (27mm)
Melting Pt: –24 °C
Density: 1.522 g/cm³
MDL Number: MFCD00000066
CAS Number: 19614-16-5
EINECS: 243-183-4
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 145-146°C/2
Melting point -24°C
Density 1.522
ND20 1.633


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR