3-Bromothioanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromophenyl methyl sulphide, 1-Bromo-3-(methylthio)benzene

004190-25G 004190-100G
101786-918EA 135.38 USD
101786-918 101786-920
3-Bromothioanisole ≥98%
3-Bromothioanisole

Hazard code(s): Irritant, Stench

Formula: C₇H₇BrS
MW: 203.1 g/mol
Boiling Pt: 124…125 °C (10mm)
MDL Number: MFCD00041395
CAS Number: 33733-73-2
UN: 3334
ADR: 9,

Order Now

Specification Test Results

Boiling point 124-125°C/1

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR