(Methylthio)acetic acid ≥98%

Supplier: Matrix Scientific

004187-5G 004187-25G
101786-912EA 66.15 USD
101786-912 101786-914
(Methylthio)acetic acid ≥98%
(Methylthio)acetic acid

Hazard code(s): Irritant, Stench

Formula: C₃H₆O₂S
Boiling Pt: 226 °C (1013 hPa)
Melting Pt: 13 - 14 °C
Density: 1.227 g/cm³ (20 °C)
MDL Number: MFCD00075444
CAS Number: 2444-37-3
EINECS: 219-483-6

Order Now

Specification Test Results

Boiling point 130-131°C/2
Melting point 13-14°C
Density 1.219
ND20 1.495


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR