3-Fluorocinnamic acid ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 3-(3-Fluorophenyl)acrylic acid, m-Fluorocinnamic acid, 3-Fluorocinnamic Acid

004180-25G 004180-100G
101786-898EA 38.46 USD
101786-898 101786-900
3-Fluorocinnamic acid ≥98%
3-Fluorocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Melting Pt: 166…167 °C
MDL Number: MFCD00004383
CAS Number: 458-46-8
EINECS: 243-902-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 166-167°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR