2-Fluorocinnamic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 3-(2-Fluorophenyl)propenoic acid

004179-25G 004179-100G
101786-892EA 29.23 USD
101786-892 101786-894
2-Fluorocinnamic acid ≥98%
2-Fluorocinnamic acid

Hazard code(s): Irritant

Formula: C₉H₇FO₂
MW: 166.15 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 178…180 °C
MDL Number: MFCD00004370
CAS Number: 451-69-4
EINECS: 207-195-3

Order Now

Specification Test Results

Melting point 178-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR