4-Fluorocinnamic acid ≥98%
Supplier: Matrix Scientific
Synonyms:
(2E)-3-(4-fluorophenyl)prop-2-enoic acid, p-Fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, trans-4-Fluorocinnamic acid
004178-25G
004178-100G
101786-888EA
21.54
USD
101786-888
101786-890
4-Fluorocinnamic acid ≥98%
4-Fluorocinnamic acid
Hazard code(s): Irritant
Formula:
C₉H₇FO₂ MW: 166.15 g/mol Melting Pt: 209…210 °C Density: 1 g/cm³ (20 °C) |
MDL Number:
MFCD00004395 CAS Number: 459-32-5 EINECS: 207-288-9 |
Specification Test Results
Melting point | 209-210°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...