2,4-Dimethylthiophenol ≥95%

Supplier: Matrix Scientific
Warning

004104-25G 004104-100G
101786-844EA 79.35 USD
101786-844 101786-846
2,4-Dimethylthiophenol ≥95%
2,4-Dimethylthiophenol

Hazard code(s): Toxic, Stench

Formula: C₈H₁₀S
Boiling Pt: 207 - 208 °C (1013 hPa)
Density: 1.027 g/cm³ (20 °C)
MDL Number: MFCD00010019
CAS Number: 13616-82-5
EINECS: 237-100-0
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 207-208°C
Density 1.02
ND20 1.5698


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR