2,5-Dimethylthiophenol ≥98%
Supplier: Matrix Scientific
|
Warning
|
004101-25G
004101-100G
101786-836EA
384.62
USD
101786-836
101786-838
2,5-Dimethylthiophenol ≥98%
2,5-Dimethylthiophenol
Hazard code(s): Toxic, Stench
Formula:
C₈H₁₀S Boiling Pt: 126 °C (50mm) Density: 1.0313 g/cm³ (20 °C) |
MDL Number:
MFCD00010020 CAS Number: 4001-61-0 EINECS: 223-649-3 UN: 2810 ADR: 6.1,III |
Specification Test Results
Boiling point | 126°C/50mm |
Density | 1.02 |
ND20 | 1.5698 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...