4,5-Dimethylthiazole ≥97%

Supplier: Matrix Scientific

004098-5G 004098-25G 004098-100G
101786-822EA 20 USD
101786-822 101786-824 101786-826
4,5-Dimethylthiazole ≥97%
4,5-Dimethylthiazole

Hazard code(s): Irritant

Formula: C₅H₇NS
MW: 113.18 g/mol
Boiling Pt: 158 °C (742mm)
Density: 1.0828 g/cm³ (20 °C)
MDL Number: MFCD00005336
CAS Number: 3581-91-7
EINECS: 222-703-3

Order Now

Specification Test Results

Boiling point 158°C/742mm
Density 1.07
ND20 1.521

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR