2,4-Dimethylthiazole ≥97%

Supplier: Matrix Scientific

004097-5G 004097-25G 004097-100G
101786-816EA 12.31 USD
101786-816 101786-818 101786-820
2,4-Dimethylthiazole ≥97%
2,4-Dimethylthiazole

Hazard code(s): Irritant

Formula: C₅H₇NS
MW: 113.18 g/mol
Boiling Pt: 145…147 °C
Density: 1.0601 g/cm³ (15 °C)
MDL Number: MFCD00014509
CAS Number: 541-58-2
EINECS: 208-786-9

Order Now

Specification Test Results

Boiling point 145-147°C
Density 1.057
ND20 1.5091

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR