2,5-Dimethylbenzylamine ≥98%

Supplier: Matrix Scientific

004075-1G 004075-5G
101786-794EA 33.85 USD
101786-794 101786-796
2,5-Dimethylbenzylamine ≥98%
2,5-Dimethylbenzylamine

Hazard code(s): Irritant, Air Sensitive

Formula: C₉H₁₃N
Boiling Pt: 121 °C (30mm)
Density: 0.95 g/cm³
MDL Number: MFCD00039780
CAS Number: 93-48-1
EINECS: 202-250-8

Order Now

Specification Test Results

Boiling point 121°C/30mm
Density 0.95


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR