3,5-Dimethylbenzylamine ≥98%

Supplier: Matrix Scientific

004072-1G 004072-5G
101786-784EA 133.85 USD
101786-784 101786-786
3,5-Dimethylbenzylamine ≥98%
3,5-Dimethylbenzylamine

Hazard code(s): Irritant, Air Sensitive

Formula: C₉H₁₃N
MW: 135.21 g/mol
Boiling Pt: 110 °C (18mm)
Density: 0.94 g/cm³
MDL Number: MFCD00040753
CAS Number: 78710-55-1
EINECS: 278-974-3

Order Now

Specification Test Results

Boiling point 110°C/18mm
Density 0.94

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR