4-Chloromethyl-m-xylene ≥98%

Supplier: Matrix Scientific

004068-5G 004068-25G
101786-772EA 90.77 USD
101786-772 101786-774
4-Chloromethyl-m-xylene ≥98%
4-Chloromethyl-m-xylene

Hazard code(s): Irritant, Lachrymator

Formula: C₉H₁₁Cl
Boiling Pt: 103 °C (15mm)
Density: 1.058 g/cm³ (19 °C)
MDL Number: MFCD00015556
CAS Number: 824-55-5
EINECS: 212-531-7
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Boiling point 103°C/15mm
Density 1.05


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR