3,5-Dimethylbenzonitrile ≥98%

Supplier: Matrix Scientific

004065-5G 004065-25G
101786-764EA 29.23 USD
101786-764 101786-766
3,5-Dimethylbenzonitrile ≥98%
3,5-Dimethylbenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₉H₉N
MW: 131.18 g/mol
Melting Pt: 42…43 °C
MDL Number: MFCD00060670
CAS Number: 22445-42-7
EINECS: 245-001-9

Order Now

Specification Test Results

Melting point 42-43°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR