2,6-Dimethylbenzonitrile ≥97%

Supplier: Matrix Scientific

004064-5G 004064-25G
101786-760EA 43.08 USD
101786-760 101786-762
2,6-Dimethylbenzonitrile ≥97%
2,6-Dimethylbenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₉H₉N
MW: 131.18 g/mol
Melting Pt: 87…89 °C
MDL Number: MFCD00037166
CAS Number: 6575-13-9
EINECS: 229-503-5

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR