2,4-Dimethylbenzonitrile

Supplier: Matrix Scientific

004062-5G 004062-25G
101786-752EA 104.62 USD
101786-752 101786-754
2,4-Dimethylbenzonitrile
2,4-Dimethylbenzonitrile

Hazard code(s): Irritant-Harmful, Refrigerate

Formula: C₉H₉N
Melting Pt: 23…25 °C
MDL Number: MFCD00016377
CAS Number: 21789-36-6
EINECS: 244-580-5

Order Now

Specification Test Results

Melting point 23-25°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR