3,5-Dimethyl-4-iodoisoxazole ≥97%

Supplier: Matrix Scientific

004051-5G 004051-25G
101786-730EA 15.38 USD
101786-730 101786-732
3,5-Dimethyl-4-iodoisoxazole ≥97%
3,5-Dimethyl-4-iodoisoxazole

Hazard code(s): Light Sensitive, Store in Freezer

Formula: C₅H₆INO
MW: 223.01 g/mol
Melting Pt: 53…55 °C
MDL Number: MFCD00173743
CAS Number: 10557-85-4

Order Now

Specification Test Results

Melting point 53-55°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR