3-Fluorophthalic anhydride ≥97% for synthesis

Supplier: Matrix Scientific

Synonyms: 4-Fluoroisobenzofuran-1,3-dione

004030-5G 004030-25G 004030-100G
101786-706EA 135.38 USD
101786-706 101786-708 101786-710
3-Fluorophthalic anhydride ≥97% for synthesis
3-Fluorophthalic anhydride

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₈H₃FO₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 157…160 °C
MDL Number: MFCD00039696
CAS Number: 652-39-1
EINECS: 211-491-8

Order Now

Specification Test Results

Melting point 157-160°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR