(3,5-Dimethoxyphenyl)acetonitrile ≥99%

Supplier: Matrix Scientific

Synonyms: 3,5-Dimethoxyphenylacetonitrile, 3,5-Dimethoxybenzyl cyanide, 2-(3,5-Dimethoxyphenyl)acetonitrile

004020-1G 004020-5G
101786-686EA 44.62 USD
101786-686 101786-688
(3,5-Dimethoxyphenyl)acetonitrile ≥99%
(3,5-Dimethoxyphenyl)acetonitrile

Hazard code(s): Irritant-Harmful

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 52…54 °C
MDL Number: MFCD00016395
CAS Number: 13388-75-5

Order Now

Specification Test Results

Melting point 52-54°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR