(2,5-Dimethoxyphenyl)acetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Dimethoxybenzyl cyanide

004019-1G 004019-5G
101786-682EA 92.31 USD
101786-682 101786-684
(2,5-Dimethoxyphenyl)acetonitrile ≥98%
(2,5-Dimethoxyphenyl)acetonitrile

Hazard code(s): Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 51…53 °C
MDL Number: MFCD00016388
CAS Number: 18086-24-3

Order Now

Specification Test Results

Melting point 51-53°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR