(3,5-Dimethoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific
Warning

004018-1G 004018-5G 004018-25G
101786-676EA 27.69 USD
101786-676 101786-678 101786-680
(3,5-Dimethoxyphenyl)acetic acid ≥98%
(3,5-Dimethoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Melting Pt: 103…105 °C
MDL Number: MFCD00016827
CAS Number: 4670-10-4

Order Now

Specification Test Results

Melting point 103-105°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR