(2,3-Dimethoxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

004017-1G 004017-5G
101786-672EA 90.77 USD
101786-672 101786-674
(2,3-Dimethoxyphenyl)acetic acid ≥98%
(2,3-Dimethoxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: (C₆H₃(OCH₃)₂)CH₂COOH
Melting Pt: 82…84 °C
MDL Number: MFCD00060351
CAS Number: 90-53-9

Order Now

Specification Test Results

Melting point 82-84°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR