3,4-Dimethoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

004015-5G 004015-25G 004015-100G
101786-662EA 21.54 USD
101786-662 101786-664 101786-666
3,4-Dimethoxyphenylboronic acid ≥97%
3,4-Dimethoxyphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₁₁BO₄
Melting Pt: 238…240 °C
MDL Number: MFCD01074574
CAS Number: 122775-35-3

Order Now

Specification Test Results

Melting point 238-240°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR