2,6-Dimethoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

004014-5G 004014-25G
101786-658EA 18.46 USD
101786-658 101786-660
2,6-Dimethoxyphenylboronic acid ≥97%
2,6-Dimethoxyphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR