2,5-Dimethoxyphenylboronic acid

Supplier: Matrix Scientific

Synonyms: (2,5-Dimethoxyphenyl)metaboric acid, 2,5-Dimethoxybenzeneboronic acid, (2,5-Dimethoxyphenyl)boronic acid, Dihydroxy(2,5-dimethoxyphenyl)borane

004013-1G 004013-5G 004013-25G
101786-652EA 21.54 USD
101786-652 101786-654 101786-656
2,5-Dimethoxyphenylboronic acid
2,5-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318181
CAS Number: 107099-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR