1,3-Dimethoxy-4-iodobenzene ≥99%

Supplier: Matrix Scientific

Synonyms: 1-Iodo-2,4-dimethoxybenzene, 2,4-Dimethoxyiodobenzene

004005-5G 004005-25G
101786-638EA 47.69 USD
101786-638 101786-640
1,3-Dimethoxy-4-iodobenzene ≥99%
1,3-Dimethoxy-4-iodobenzene

Hazard code(s): Irritant, Light Sensitive

Formula: C₈H₉IO₂
MW: 264.06 g/mol
Melting Pt: 37…39 °C
MDL Number: MFCD00059268
CAS Number: 20469-63-0

Order Now

Specification Test Results

Melting point 37-39°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR