5,6-Dimethoxyindole ≥98%

Supplier: Matrix Scientific

004004-100MG 004004-500MG
101786-634EA 53.85 USD
101786-634 101786-636
5,6-Dimethoxyindole ≥98%
5,6-Dimethoxyindole

Hazard code(s): Irritant

Formula: C₁₀H₁₁NO₂
MW: 177.2 g/mol
Melting Pt: 157…159 °C
MDL Number: MFCD00005675
CAS Number: 14430-23-0
EINECS: 238-402-5

Order Now

Specification Test Results

Melting point 157-159°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR