Veratrohydrazide ≥98%

Supplier: Matrix Scientific

Synonyms: 3,4-Dimethoxybenzohydrazide, 3,4-Dimethoxybenzene-1-carbohydrazide, 3,4-Dimethoxybenzoic hydrazide, 3,4-Dimethoxybenzoic acid hydrazide, 3,4-Dimethoxybenzhydrazide, 3,4-Dimethoxybenzenecarbohydrazide

003997-5G 003997-25G
101786-614EA 21.54 USD
101786-614 101786-616
Veratrohydrazide ≥98%
Veratrohydrazide

Hazard code(s): Irritant

Formula: C₉H₁₂N₂O₃
MW: 196.21 g/mol
Melting Pt: 145…146 °C
MDL Number: MFCD00017061
CAS Number: 41764-74-3
EINECS: 255-541-7

Order Now

Specification Test Results

Melting point 145-146°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR